Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate

Lieferant: Apollo Scientific
Achtung

OR15226-1G
APOSOR15226-1GEA 0 EUR
APOSOR15226-1G
Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate
Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate
Formel: C₁₁H₁₂N₂O₃
Molecular Weight: 220,23 g/mol
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443989
CAS-Nummer: 627901-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR