1-Boc-4-(2-chloroethyl)piperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Chloroethyl)piperazine,N1-BOC protected, tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

OR15245-1G
APOSOR15245-1GEA 21.9 EUR
APOSOR15245-1G
1-Boc-4-(2-chloroethyl)piperazine
1-Boc-4-(2-chloroethyl)piperazine
Formel: C₁₁H₂₁ClN₂O₂
Molecular Weight: 248,751 g/mol
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443993
CAS-Nummer: 208167-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR