5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%

Lieferant: Apollo Scientific
Achtung

OR15246-1G
APOSOR15246-1GEA 91.5 EUR
APOSOR15246-1G
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde
Formel: C₁₂H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06246086
CAS-Nummer: 85943-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR