1-(2-Amino-3-pyridinyl)-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Aminopyridin-3-yl)ethanone, 3-Acetyl-2-aminopyridine

OR15249-5G
APOSOR15249-5GEA 66.7 EUR
APOSOR15249-5G
1-(2-Amino-3-pyridinyl)-1-ethanone
1-(2-Amino-3-pyridinyl)-1-ethanone
Formel: C₇H₈N₂O
Molecular Weight: 136.15 g/mol
Schmelzpunkt: 139…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258895
CAS-Nummer: 65326-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR