4-Bromo-1H-pyrrole-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR15253-1G
APOSOR15253-1GEA 21.9 EUR
APOSOR15253-1G
4-Bromo-1H-pyrrole-2-carboxylic acid 97%
4-Bromo-1H-pyrrole-2-carboxylic acid
Formel: C₅H₄BrNO₂
Molecular Weight: 189.9948 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00832862
CAS-Nummer: 27746-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR