2-(Chloromethyl)-1-methyl-1H-imidazole

Lieferant: Apollo Scientific
Achtung

OR15254-100MG
APOSOR15254-100MGEA 21.9 EUR
APOSOR15254-100MG
2-(Chloromethyl)-1-methyl-1H-imidazole
2-(Chloromethyl)-1-methyl-1H-imidazole
Formel: C₅H₇ClN₂
Molecular Weight: 130.58 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234119
CAS-Nummer: 19225-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR