4-Chlor-1H-pyrrolo[2,3-b]pyridin
Lieferant: Apollo Scientific
|
Gefahr
|
OR15256-5G
APOSOR15256-5GEA
24.8
EUR
APOSOR15256-5G
4-Chlor-1H-pyrrolo[2,3-b]pyridin
4-Chloro-7-azaindole
Formel:
C₇H₅ClN₂ Schmelzpunkt: 176…181 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08272232 CAS-Nummer: 55052-28-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...