2-Amino-5-bromo-3-(hydroxymethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-bromo-3-pyridinemethanol

OR15266-5G
APOSOR15266-5GEA 55.1 EUR
APOSOR15266-5G
2-Amino-5-bromo-3-(hydroxymethyl)pyridine
2-Amino-5-bromo-3-(hydroxymethyl)pyridine
Formel: C₆H₇BrN₂O
Molecular Weight: 203.039 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05664025
CAS-Nummer: 335031-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR