6-[(tert-Butoxycarbonyl)amino]nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Aminonicotinic acid, 6-BOC protected, 6-((tert-Butoxycarbonyl)amino)nicotinic acid
OR15274-1G
OR15274-250MG
APOSOR15274-1GEA
130
EUR
APOSOR15274-1G
APOSOR15274-250MG
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
Formel:
C₁₁H₁₄N₂O₄ Molecular Weight: 238.25 g/mol Schmelzpunkt: 292…296 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682388 CAS-Nummer: 231958-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...