(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine

OR15278-1G OR15278-5G
APOSOR15278-1GEA 336 EUR
APOSOR15278-1G APOSOR15278-5G
(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine
(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine
Formel: C₁₀H₁₂ClN₃
Molecular Weight: 209.68 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06496216
CAS-Nummer: 866142-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR