(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine
OR15278-1G
OR15278-5G
APOSOR15278-1GEA
336
EUR
APOSOR15278-1G
APOSOR15278-5G
(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine
(6-Chloroimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine
Formel:
C₁₀H₁₂ClN₃ Molecular Weight: 209.68 g/mol Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06496216 CAS-Nummer: 866142-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...