Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

OR1528-1G
APOSOR1528-1GEA 0 EUR
APOSOR1528-1G
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Formel: C₁₂H₁₀ClNO₃S
Molecular Weight: 283,74 g/mol
Schmelzpunkt: 137…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00816787
CAS-Nummer: 263016-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR