1-(5-Chloro-2-nitrophenyl)-1-ethanone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5'-Chloro-2'-nitroacetophenone, 1-(5-Chloro-2-nitrophenyl)ethanone
OR15281-1G
APOSOR15281-1GEA
21.9
EUR
APOSOR15281-1G
1-(5-Chloro-2-nitrophenyl)-1-ethanone
1-(5-Chloro-2-nitrophenyl)-1-ethanone
Formel:
C₈H₆ClNO₃ Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787322 CAS-Nummer: 18640-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...