Ethyl-1-(methylsulphonyl)-4-piperidinecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR15286-1G
APOSOR15286-1GEA
0
EUR
APOSOR15286-1G
Ethyl-1-(methylsulphonyl)-4-piperidinecarboxylate
Ethyl-1-(methylsulphonyl)-4-piperidinecarboxylate
Formel:
C₉H₁₇NO₄S Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00115615 CAS-Nummer: 217487-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...