tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Lieferant: Apollo Scientific

Synonyme: 4-(3-Acetyl-4-nitrophenyl)piperazine, N1-BOC protected

OR15299-500MG
APOSOR15299-500MGEA 306 EUR
APOSOR15299-500MG
tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₇H₂₃N₃O₅
Molecular Weight: 349,39 g/mol
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276930
CAS-Nummer: 886361-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR