1-(2-Amino-5-morpholinophenyl)-1-ethanone 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR15306-500MG
APOSOR15306-500MGEA
306
EUR
APOSOR15306-500MG
1-(2-Amino-5-morpholinophenyl)-1-ethanone 95%
1-(2-Amino-5-morpholinophenyl)-1-ethanone
Formel:
C₁₂H₁₆N₂O₂ Molecular Weight: 220,27 g/mol Schmelzpunkt: 104…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787743 CAS-Nummer: 98440-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...