2-Amino-3-brom-5-nitrobenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR1531-100G
APOSOR1531-100GEA
95.7
EUR
APOSOR1531-100G
2-Amino-3-brom-5-nitrobenzonitril 98%
2-Amino-3-brom-5-nitrobenzonitril
Formel:
C₇H₄BrN₃O₂ Molecular Weight: 242,03 g/mol Schmelzpunkt: 180…185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00054185 CAS-Nummer: 17601-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...