5-Bromo-2-hydrazino-1H-1,3-benzimidazole 95%

Lieferant: Apollo Scientific
Achtung

OR15310-1G
APOSOR15310-1GEA 336 EUR
APOSOR15310-1G
5-Bromo-2-hydrazino-1H-1,3-benzimidazole 95%
5-Bromo-2-hydrazino-1H-1,3-benzimidazole
Formel: C₇H₇BrN₄
Molecular Weight: 227,07 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276936
CAS-Nummer: 54303-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR