4-Bromo-1-methyl-1H-pyrrole-2-carbohydrazide
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid hydrazide
OR15311-500MG
APOSOR15311-500MGEA
336
EUR
APOSOR15311-500MG
4-Bromo-1-methyl-1H-pyrrole-2-carbohydrazide
4-Bromo-1-methyl-1H-pyrrole-2-carbohydrazide
Formel:
C₆H₈BrN₃O Molecular Weight: 218,06 g/mol Schmelzpunkt: 166…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07838416 CAS-Nummer: 875163-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...