4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid

Lieferant: Apollo Scientific

OR15322-1G
APOSOR15322-1GEA 538 EUR
APOSOR15322-1G
4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid
4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid
Formel: C₁₉H₁₉ClO₃
Schmelzpunkt: 154…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664417
CAS-Nummer: 344281-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR