Ethyl-2-(4-methoxyphenyl)-1,3-thiazol-4-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(4-methoxyphenyl)thiazole-4-carboxylate

OR15332-1G
APOSOR15332-1GEA 29 EUR
APOSOR15332-1G
Ethyl-2-(4-methoxyphenyl)-1,3-thiazol-4-carboxylat
Ethyl-2-(4-methoxyphenyl)-1,3-thiazol-4-carboxylat
Formel: C₁₃H₁₃NO₃S
Schmelzpunkt: 94…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738343
CAS-Nummer: 57677-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR