Methyl-4-chlor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR15352-10G
OR15352-1G
OR15352-5G
APOSOR15352-10GEA
0
EUR
APOSOR15352-10G
APOSOR15352-1G
APOSOR15352-5G
Methyl-4-chlor-2-benzo[b]thiophencarboxylat
Methyl-4-chlor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇ClO₂S Schmelzpunkt: 95…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371546 CAS-Nummer: 35212-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...