2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Oxo-1,3-oxazolidin-3-yl)ethyl toluene-4-sulphonate

OR15356-1G
APOSOR15356-1GEA 59.6 EUR
APOSOR15356-1G
2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulphonate
2-(2-Oxo-1,3-oxazolan-3-yl)ethyl4-methylbenzenesulphonate
Formel: C₁₂H₁₅NO₅S
Molecular Weight: 285,32 g/mol
Schmelzpunkt: 68…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276953
CAS-Nummer: 159974-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR