(3-Phenyl-5-isoxazolyl)methanamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Aminomethyl)-3-phenylisoxazole, (3-Phenylisoxazol-5-yl)methanamine

OR15367-1G
APOSOR15367-1GEA 0 EUR
APOSOR15367-1G
(3-Phenyl-5-isoxazolyl)methanamine 97%
(3-Phenyl-5-isoxazolyl)methanamine
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,2 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03844852
CAS-Nummer: 54408-35-4
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR