2-(Thien-2-yl)pyrimidine-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Carboxypyrimidin-2-yl)thiophene, 2-(Thiophen-2-yl)pyrimidine-5-carboxylic acid, 5-Carboxy-2-(thien-2-yl)pyrimidine, 5-Carboxy-2-(thien-2-yl)-1,3-diazine

OR15373-1G
APOSOR15373-1GEA 255 EUR
APOSOR15373-1G
2-(Thien-2-yl)pyrimidine-5-carboxylic acid 97%
2-(Thien-2-yl)pyrimidine-5-carboxylic acid
Formel: C₉H₆N₂O₂S
Molecular Weight: 206,221 g/mol
Schmelzpunkt: 291…296 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817551
CAS-Nummer: 916766-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR