3-Phenylisoxazole-5-carbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR15378-1G
APOSOR15378-1GEA 166 EUR
APOSOR15378-1G
3-Phenylisoxazole-5-carbaldehyde 97%
3-Phenylisoxazole-5-carbaldehyde
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
Schmelzpunkt: 72,5…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817543
CAS-Nummer: 72418-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR