tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile, N6-BOC protected

OR15388-1G
APOSOR15388-1GEA 30.6 EUR
APOSOR15388-1G
tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate
tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate
Formel: C₁₃H₁₇N₃O₂S
Schmelzpunkt: 193…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664037
CAS-Nummer: 150986-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR