6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-tert-Butyl 3-methyl 2-amino-4,5-dihydrothieno[2,3-c]pyridine-3,6(7H)-dicarboxylate, Methyl 2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate, N6-BOC protected

OR15389-1G
APOSOR15389-1GEA 24.8 EUR
APOSOR15389-1G
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Formel: C₁₄H₂₀N₂O₄S
Molecular Weight: 312,39 g/mol
Schmelzpunkt: 165…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200952
CAS-Nummer: 877041-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR