2-Chloro-N-(2-furylmethyl)acetamide

Lieferant: Apollo Scientific
Achtung

OR15392-1G
APOSOR15392-1GEA 58 EUR
APOSOR15392-1G
2-Chloro-N-(2-furylmethyl)acetamide
2-Chloro-N-(2-furylmethyl)acetamide
Formel: C₇H₈ClNO₂
Molecular Weight: 173,6 g/mol
Schmelzpunkt: 39…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00297044
CAS-Nummer: 40914-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR