Ethyl-2-cyano-3-(4-methylphenyl)acrylate
Lieferant: Apollo Scientific
|
Achtung
|
OR15405-1G
APOSOR15405-1GEA
34.8
EUR
APOSOR15405-1G
Ethyl-2-cyano-3-(4-methylphenyl)acrylate
Ethyl-2-cyano-3-(4-methylphenyl)acrylate
Formel:
C₁₃H₁₃NO₂ Schmelzpunkt: 86…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136329 CAS-Nummer: 2017-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...