Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate
Lieferant: Apollo Scientific
OR15409-1G
APOSOR15409-1GEA
0
EUR
APOSOR15409-1G
Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate
Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate
Formel:
C₁₃H₁₇N₃O₄ Molecular Weight: 279,3 g/mol Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01308911 CAS-Nummer: 300804-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...