Methyl-2-amino-3-methylbenzoat 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-3-methylbenzoate

OR15415-1G
APOSOR15415-1GEA 21.9 EUR
APOSOR15415-1G
Methyl-2-amino-3-methylbenzoat 96%
Methyl-2-amino-3-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06200918
CAS-Nummer: 22223-49-0
EINECS: 244-847-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR