Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR15417-1G
APOSOR15417-1GEA
40.6
EUR
APOSOR15417-1G
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate
Formel:
C₁₀H₁₁N₅O₂ Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00838109 CAS-Nummer: 23947-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...