Methyl-3-phenyl-1H-pyrazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Methoxycarbonyl)-3-phenyl-1H-pyrazole

OR15423-1G
APOSOR15423-1GEA 0 EUR
APOSOR15423-1G
Methyl-3-phenyl-1H-pyrazole-5-carboxylate
Methyl-3-phenyl-1H-pyrazole-5-carboxylate
Formel: C₁₁H₁₀N₂O₂
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00208415
CAS-Nummer: 856061-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR