5-[(4-Methylphenyl)sulphonyl]-2-phenyl-4-pyrimidinol

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-5-[(4-methylphenyl)sulphonyl]-2-phenylpyrimidine

OR15424-10G OR15424-1G OR15424-5G
APOSOR15424-10GEA 1690 EUR
APOSOR15424-10G APOSOR15424-1G APOSOR15424-5G
5-[(4-Methylphenyl)sulphonyl]-2-phenyl-4-pyrimidinol
5-[(4-Methylphenyl)sulphonyl]-2-phenyl-4-pyrimidinol
Formel: C₁₇H₁₄N₂O₃S
Molecular Weight: 326.38 g/mol
Schmelzpunkt: 325…328 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200982
CAS-Nummer: 885949-45-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR