2-Phenyl-1,3-thiazole-4-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR15428-1G
APOSOR15428-1GEA 109 EUR
APOSOR15428-1G
2-Phenyl-1,3-thiazole-4-carbohydrazide
2-Phenyl-1,3-thiazole-4-carbohydrazide
Formel: C₁₀H₉N₃OS
Molecular Weight: 219.27 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00174011
CAS-Nummer: 7113-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR