4'-(Methylsulfonyl)acetophenon 98%

Lieferant: Apollo Scientific

OR1543-25G
APOSOR1543-25GEA 0 EUR
APOSOR1543-25G
4'-(Methylsulfonyl)acetophenon 98%
4'-(Methylsulfonyl)acetophenon
Formel: C₉H₁₀O₃S
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025072
CAS-Nummer: 10297-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR