1-{[4-(Acetylamino)phenyl]sulphonyl}-4-piperidinecarboxylic acid 95%

Lieferant: Apollo Scientific

OR15431-1G
APOSOR15431-1GEA 127 EUR
APOSOR15431-1G
1-{[4-(Acetylamino)phenyl]sulphonyl}-4-piperidinecarboxylic acid 95%
1-{[4-(Acetylamino)phenyl]sulphonyl}-4-piperidinecarboxylic acid
Formel: C₁₄H₁₈N₂O₅S
Molecular Weight: 326,37 g/mol
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02081043
CAS-Nummer: 314744-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR