[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl]methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR15434-500MG
APOSOR15434-500MGEA
121
EUR
APOSOR15434-500MG
[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl]methanol
[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl]methanol
Formel:
C₉H₇ClN₂O₂ Schmelzpunkt: 96…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200919 CAS-Nummer: 5372-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...