4,5-Dibromo-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15435-250MG
APOSOR15435-250MGEA 72.5 EUR
APOSOR15435-250MG
4,5-Dibromo-1H-pyrrole-2-carboxylic acid
4,5-Dibromo-1H-pyrrole-2-carboxylic acid
Formel: C₅H₃Br₂NO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01348229
CAS-Nummer: 34649-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR