4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15439-1G
APOSOR15439-1GEA 136 EUR
APOSOR15439-1G
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
Formel: C₁₄H₁₀Cl₂O₃
Molecular Weight: 297.14 g/mol
Schmelzpunkt: 210…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200988
CAS-Nummer: 56442-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR