Ethyl-1,3-benzothiazol-2-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR15441-1G
OR15441-5G
APOSOR15441-1GEA
21.9
EUR
APOSOR15441-1G
APOSOR15441-5G
Ethyl-1,3-benzothiazol-2-carboxylat
Formel:
C₁₀H₉NO₂S Schmelzpunkt: 68…72 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00848360 CAS-Nummer: 32137-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...