Ethyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetate

Lieferant: Apollo Scientific
Achtung

OR15442-1G
APOSOR15442-1GEA 0 EUR
APOSOR15442-1G
Ethyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetate
Ethyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetate
Formel: C₁₃H₁₃NO₄
Molecular Weight: 247,25 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200915
CAS-Nummer: 14771-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR