6-Iodimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific
Achtung

OR15445-1G
APOSOR15445-1GEA 21.9 EUR
APOSOR15445-1G
6-Iodimidazo[1,2-a]pyridin
6-Iodimidazo[1,2-a]pyridin
Formel: C₇H₅IN₂
Molecular Weight: 244,0300 g/mol
Schmelzpunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200985
CAS-Nummer: 426825-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR