Methyl-3,5-diiodo-1H-indole-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR15451-1G
APOSOR15451-1GEA
0
EUR
APOSOR15451-1G
Methyl-3,5-diiodo-1H-indole-2-carboxylate
Methyl-3,5-diiodo-1H-indole-2-carboxylate
Formel:
C₁₀H₇I₂NO₂ Schmelzpunkt: 209…211 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06200993 CAS-Nummer: 885949-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...