3-(2-Methyl-1,3-dioxolan-2-yl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
OR15454-250MG
APOSOR15454-250MGEA
21.9
EUR
APOSOR15454-250MG
3-(2-Methyl-1,3-dioxolan-2-yl)aniline
3-(2-Methyl-1,3-dioxolan-2-yl)aniline
Formel:
C₁₀H₁₃NO₂ Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020904 CAS-Nummer: 51226-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...