4-Propionyl-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15460-1G
APOSOR15460-1GEA 243 EUR
APOSOR15460-1G
4-Propionyl-1H-pyrrole-2-carboxylic acid
4-Propionyl-1H-pyrrole-2-carboxylic acid
Formel: C₈H₉NO₃
Molecular Weight: 167.17 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200986
CAS-Nummer: 111468-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR