2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
Lieferant: Apollo Scientific
|
Achtung
|
OR15487-250MG
APOSOR15487-250MGEA
36.4
EUR
APOSOR15487-250MG
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
Formel:
C₈H₁₀N₂ Schmelzpunkt: 162…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00030437 CAS-Nummer: 10087-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...