1-(Piperidin-4-yl)-1H-benzotriazole hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1H-Benzotriazol-1-yl)piperidine hydrochloride

OR1549-1G
APOSOR1549-1GEA 314 EUR
APOSOR1549-1G
1-(Piperidin-4-yl)-1H-benzotriazole hydrochloride
1-(Piperidin-4-yl)-1H-benzotriazole hydrochloride
Formel: C₁₁H₁₄N₄·ClH
Schmelzpunkt: 314…315 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00831110
CAS-Nummer: 79098-80-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR