1-(2-Furyl)-2,2-dimethyl-1,3-propanediol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(1,3-Dihydroxy-2,2-dimethylprop-1-yl)furan

OR15495-500MG
APOSOR15495-500MGEA 560 EUR
APOSOR15495-500MG
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00720174
CAS-Nummer: 4946-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR